Search
Search

HSP90AA1| Gene Symbol: HSP90AA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066239.1 | XP_017921728.1 | 7 - 8 | 1492 | 108 | |
| GenBank mRNA | AF548366.1 | - | 4 - 5 | 752 | 108 | |
| JO581047.1 | - | 3438 | 108 | |||
| JR123019.1 | - | 1714 | 108 | |||
Set Membership: |
|