Search
Search
| Gene Symbol: RAB17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040662.1 | XP_017896151.1 | 2 - 3 | 539 | 83 | |
| XM_018040668.1 | XP_017896157.1 | 2 - 3 | 451 | 83 | ||
| XM_018040671.1 | XP_017896160.1 | 2 - 3 | 539 | 83 | ||
| GenBank mRNA | JO402301.2 | - | 4 - 5 | 1157 | 83 | |
| JO435706.2 | - | 2 - 3 | 276 | 83 | ||
Set Membership: |
|