Search
Search
| Gene Symbol: LIN7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047602.1 | XP_017903091.1 | 3 - 4 | 1154 | 79 | |
| XM_018047603.1 | XP_017903092.1 | 3 - 4 | 288 | 79 | ||
| XM_018047604.1 | XP_017903093.1 | 3 - 4 | 515 | 79 | ||
Set Membership: |
|