Search
Search
| Gene Symbol: PMEL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048106.1 | XP_017903595.1 | 10 - 11 | 2161 | 114 | |
| XM_018048107.1 | XP_017903596.1 | 10 - 11 | 2141 | 114 | ||
| XM_018048108.1 | XP_017903597.1 | 10 - 11 | 2152 | 114 | ||
Set Membership: |
|