Search
Search

ATP2C1| Gene Symbol: ATP2C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051409.1 | XP_017906898.1 | 9 - 10 | 1166 | 119 | |
| XM_018051417.1 | XP_017906906.1 | 11 - 12 | 1113 | 119 | ||
| XM_018051427.1 | XP_017906916.1 | 11 - 12 | 954 | 119 | ||
| XM_018051432.1 | XP_017906921.1 | 10 - 11 | 1049 | 119 | ||
| XM_018051436.1 | XP_017906925.1 | 9 - 10 | 924 | 119 | ||
| GenBank mRNA | JO426167.1 | - | 18 - 19 | 2309 | 119 | |
| JO644856.2 | - | 4 - 5 | 397 | 119 | ||
Set Membership: |
|