Search
Search

ARHGAP44| Gene Symbol: ARHGAP44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064675.1 | XP_017920164.1 | 8 - 9 | 921 | 62 | |
| XM_018064677.1 | XP_017920166.1 | 8 - 9 | 921 | 62 | ||
| XM_018064678.1 | XP_017920167.1 | 8 - 9 | 921 | 62 | ||
| GenBank mRNA | JO633804.2 | - | 14 - 15 | 3121 | 62 | |
Set Membership: |
|