Search
Search

TMEM63B| Gene Symbol: TMEM63B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038929.1 | XP_017894418.1 | 8 - 9 | 748 | 64 | |
| XM_018038930.1 | XP_017894419.1 | 8 - 9 | 913 | 64 | ||
| XM_018038931.1 | XP_017894420.1 | 8 - 9 | 874 | 64 | ||
| XM_018038932.1 | XP_017894421.1 | 7 - 8 | 709 | 64 | ||
| GenBank mRNA | JI861659.1 | - | 7 - 8 | 603 | 64 | |
| KA339078.1 | - | 8 - 9 | 779 | 64 | ||
Set Membership: |
|