Search
Search

TRERF1| Gene Symbol: TRERF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039088.1 | XP_017894577.1 | 12 - 13 | 3551 | 59 | |
| XM_018039089.1 | XP_017894578.1 | 13 - 14 | 3503 | 59 | ||
| XM_018039090.1 | XP_017894579.1 | 11 - 12 | 3361 | 59 | ||
| XM_018039091.1 | XP_017894580.1 | 12 - 13 | 3551 | 59 | ||
| XM_018039092.1 | XP_017894581.1 | 11 - 12 | 3305 | 59 | ||
| XM_018039093.1 | XP_017894582.1 | 11 - 12 | 3305 | 59 | ||
| XM_018039094.1 | XP_017894583.1 | 12 - 13 | 3611 | 59 | ||
Set Membership: |
|