Search
Search
| Gene Symbol: CEP83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679823.3 | XP_005679880.2 | 14 - 15 | 2358 | 158 | |
| XM_005679824.3 | XP_005679881.2 | 13 - 14 | 2109 | 158 | ||
| XM_018047628.1 | XP_017903117.1 | 15 - 16 | 2409 | 158 | ||
| GenBank mRNA | JO595519.2 | - | 2 - 3 | 563 | 158 | |
Set Membership: |
|