Search
Search

C5H12orf4| Gene Symbol: C5H12orf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680983.3 | XP_005681040.2 | 9 - 10 | 1521 | 122 | |
| XM_018048613.1 | XP_017904102.1 | 8 - 9 | 1117 | 122 | ||
| GenBank mRNA | JO608520.2 | - | 9 - 10 | 1245 | 122 | |
Set Membership: |
|