Search
Search

SEMA6C| Gene Symbol: SEMA6C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013962422.2 | XP_013817876.2 | 13 - 14 | 1619 | 63 | |
| XM_013962424.2 | XP_013817878.2 | 13 - 14 | 1620 | 63 | ||
| XM_013962425.2 | XP_013817879.2 | 7 - 8 | 873 | 63 | ||
| XM_018046064.1 | XP_017901553.1 | 11 - 12 | 2059 | 63 | ||
| XM_018046065.1 | XP_017901554.1 | 13 - 14 | 1620 | 63 | ||
Set Membership: |
|