Search
Search
| Gene Symbol: RYR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062343.1 | XP_017917832.1 | 100 - 101 | 14649 | 51 | |
| XM_018062344.1 | XP_017917833.1 | 99 - 100 | 14634 | 51 | ||
| XM_018062345.1 | XP_017917834.1 | 99 - 100 | 14544 | 51 | ||
| XM_018062346.1 | XP_017917835.1 | 98 - 99 | 14529 | 51 | ||
Set Membership: |
|