Search
Search

SLC16A6| Gene Symbol: SLC16A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063939.1 | XP_017919428.1 | 4 - 5 | 663 | 66 | |
| XM_018063940.1 | XP_017919429.1 | 5 - 6 | 791 | 66 | ||
| XM_018063941.1 | XP_017919430.1 | 4 - 5 | 846 | 66 | ||
| GenBank mRNA | JO427631.1 | - | 5 - 6 | 889 | 66 | |
| JO428131.1 | - | 4 - 5 | 760 | 66 | ||
Set Membership: |
|