Search
Search
| Gene Symbol: CD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043500.1 | XP_017898989.1 | 2 - 3 | 301 | 67 | |
| XM_018043501.1 | XP_017898990.1 | 2 - 3 | 301 | 67 | ||
| XM_018043502.1 | XP_017898991.1 | 2 - 3 | 301 | 67 | ||
| XM_018043503.1 | XP_017898992.1 | 2 - 3 | 301 | 67 | ||
| XM_018043504.1 | XP_017898993.1 | 2 - 3 | 301 | 67 | ||
| XM_018043505.1 | XP_017898994.1 | 2 - 3 | 301 | 67 | ||
| GenBank mRNA | JO612402.2 | - | 2 - 3 | 460 | 67 | |
Set Membership: |
|