Search
Search

LOC102182573| Gene Symbol: LOC102182573 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680838.3 | XP_005680895.2 | 2 - 3 | 320 | 89 | |
| GenBank mRNA | JO591048.2 | - | 5 - 6 | 496 | 89 | |
| JO591049.2 | - | 4 - 5 | 472 | 89 | ||
Set Membership: |
|