Search
Search

RNF43
LOC108634861| Gene Symbol: RNF43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064116.1 | XP_017919605.1 | 4 - 5 | 2604 | 65 | |
| XM_018064117.1 | XP_017919606.1 | 5 - 6 | 1551 | 65 | ||
| XM_018064118.1 | XP_017919607.1 | 4 - 5 | 2602 | 65 | ||
| XM_018064119.1 | XP_017919608.1 | 3 - 4 | 2775 | 65 | ||
| Gene Symbol: LOC108634861 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045209.1 | XP_017900698.1 | 3 - 4 | 276 | 65 | |
Set Membership: |
|