Search
Search

COL4A3| Gene Symbol: COL4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058855.1 | XP_017914344.1 | 22 - 23 | 1588 | 97 | |
| GenBank mRNA | JT672045.1 | - | 24 - 25 | 1867 | 97 | |
Set Membership: |
|