Search
Search
| Gene Symbol: CD86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675017.2 | XP_005675074.1 | 3 - 4 | 545 | 107 | |
| XM_005675019.3 | XP_005675076.1 | 3 - 4 | 524 | 107 | ||
| XM_018045106.1 | XP_017900595.1 | 3 - 4 | 560 | 107 | ||
| XM_018045206.1 | XP_017900695.1 | 3 - 4 | 539 | 107 | ||
| XM_018045277.1 | XP_017900766.1 | 3 - 4 | 560 | 107 | ||
| XM_018045320.1 | XP_017900809.1 | 3 - 4 | 560 | 107 | ||
| XM_018045360.1 | XP_017900849.1 | 3 - 4 | 560 | 107 | ||
| GenBank mRNA | JO319628.2 | - | 3 - 4 | 564 | 107 | |
| JO319629.2 | - | 3 - 4 | 542 | 107 | ||
| JO319630.2 | - | 3 - 4 | 521 | 107 | ||
| JO319631.2 | - | 3 - 4 | 542 | 107 | ||
| JO319632.2 | - | 3 - 4 | 585 | 107 | ||
| JO319633.2 | - | 3 - 4 | 564 | 107 | ||
| JO319634.2 | - | 3 - 4 | 585 | 107 | ||
| JO319636.2 | - | 3 - 4 | 521 | 107 | ||
| JO361468.2 | - | 2 - 3 | 437 | 107 | ||
| JO419205.2 | - | 1 - 2 | 213 | 107 | ||
| JO552365.2 | - | 1 - 2 | 106 | 107 | ||
| JO561181.2 | - | 3 - 4 | 552 | 107 | ||
| JO562913.2 | - | 3 - 4 | 552 | 107 | ||
| JO566731.2 | - | 3 - 4 | 552 | 107 | ||
| JO577611.1 | - | 3 - 4 | 641 | 107 | ||
| JO577612.1 | - | 3 - 4 | 610 | 107 | ||
| JO577613.1 | - | 3 - 4 | 641 | 107 | ||
| JO577614.1 | - | 3 - 4 | 641 | 107 | ||
| JT717513.1 | - | 3 - 4 | 496 | 107 | ||
Set Membership: |
|