Search
Search

LETMD1| Gene Symbol: LETMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679960.3 | XP_005680017.1 | 7 - 8 | 1015 | 63 | |
| XM_013963804.2 | XP_013819258.1 | 4 - 5 | 643 | 63 | ||
Set Membership: |
|