Search
Search
| Gene Symbol: CBLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013966815.2 | XP_013822269.1 | 1 - 2 | 1078 | 64 | |
| XM_013966816.2 | XP_013822270.1 | 1 - 2 | 1078 | 64 | ||
| XM_013966818.2 | XP_013822272.1 | 1 - 2 | 1078 | 64 | ||
| XM_013966819.2 | XP_013822273.1 | 1 - 2 | 1078 | 64 | ||
| XM_018054655.1 | XP_017910144.1 | 1 - 2 | 1078 | 64 | ||
Set Membership: |
|