Search
Search
| Gene Symbol: DYM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039567.1 | XP_017895056.1 | 8 - 9 | 1060 | 63 | |
| XM_018039568.1 | XP_017895057.1 | 8 - 9 | 1056 | 63 | ||
| XM_018039569.1 | XP_017895058.1 | 8 - 9 | 1061 | 63 | ||
Set Membership: |
|