Search
Search
| Gene Symbol: BANP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061833.1 | XP_017917322.1 | 10 - 11 | 1812 | 62 | |
| XM_018061834.1 | XP_017917323.1 | 10 - 11 | 1803 | 62 | ||
| XM_018061835.1 | XP_017917324.1 | 10 - 11 | 1809 | 62 | ||
| XM_018061836.1 | XP_017917325.1 | 10 - 11 | 1800 | 62 | ||
| XM_018061837.1 | XP_017917326.1 | 9 - 10 | 1694 | 62 | ||
| XM_018061838.1 | XP_017917327.1 | 9 - 10 | 1685 | 62 | ||
| XM_018061839.1 | XP_017917328.1 | 9 - 10 | 1682 | 62 | ||
Set Membership: |
|