Search
Search

LOC102175560| Gene Symbol: LOC102175560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680884.3 | XP_005680941.2 | 11 - 12 | 1560 | 84 | |
| XM_018048527.1 | XP_017904016.1 | 11 - 12 | 1347 | 84 | ||
Set Membership: |
|