Search
Search
| Gene Symbol: NIPBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065528.1 | XP_017921017.1 | 41 - 42 | 7622 | 96 | |
| XM_018065529.1 | XP_017921018.1 | 42 - 43 | 7516 | 96 | ||
| XM_018065530.1 | XP_017921019.1 | 41 - 42 | 7451 | 96 | ||
| XM_018065531.1 | XP_017921020.1 | 41 - 42 | 7478 | 96 | ||
| XM_018065532.1 | XP_017921021.1 | 40 - 41 | 5852 | 96 | ||
Set Membership: |
|