Search
Search

CCDC114| Gene Symbol: CCDC114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062696.1 | XP_017918185.1 | 11 - 12 | 1981 | 101 | |
| XM_018062697.1 | XP_017918186.1 | 12 - 13 | 4218 | 101 | ||
| XM_018062698.1 | XP_017918187.1 | 12 - 13 | 1672 | 101 | ||
| XM_018062699.1 | XP_017918188.1 | 10 - 11 | 1420 | 101 | ||
| XM_018062700.1 | XP_017918189.1 | 5 - 6 | 1131 | 101 | ||
| GenBank mRNA | JO591065.2 | - | 3 - 4 | 898 | 101 | |
Set Membership: |
|