Search
Search

ZNF446| Gene Symbol: ZNF446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063547.1 | XP_017919036.1 | 3 - 4 | 1569 | 78 | |
| XM_018063548.1 | XP_017919037.1 | 4 - 5 | 1168 | 78 | ||
| XM_018063549.1 | XP_017919038.1 | 4 - 5 | 1818 | 78 | ||
| XM_018063550.1 | XP_017919039.1 | 3 - 4 | 1533 | 78 | ||
Set Membership: |
|