Search
Search

ATP11B| Gene Symbol: ATP11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675219.3 | XP_005675276.1 | 7 - 8 | 640 | 78 | |
| XM_018047490.1 | XP_017902979.1 | 9 - 10 | 1057 | 78 | ||
| XM_018047495.1 | XP_017902984.1 | 9 - 10 | 1059 | 78 | ||
Set Membership: |
|