Search
Search
| Gene Symbol: ANXA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055460.1 | XP_017910949.1 | 3 - 4 | 264 | 63 | |
| XM_018055461.1 | XP_017910950.1 | 3 - 4 | 165 | 63 | ||
| GenBank mRNA | JO433293.2 | - | 3 - 4 | 307 | 63 | |
| JO576500.1 | - | 10 - 11 | 1373 | 63 | ||
Set Membership: |
|