Search
Search

ZNF287| Gene Symbol: ZNF287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064693.1 | XP_017920182.1 | 4 - 5 | 1053 | 87 | |
| XM_018064694.1 | XP_017920183.1 | 5 - 6 | 1113 | 87 | ||
| XM_018064695.1 | XP_017920184.1 | 2 - 3 | 730 | 87 | ||
Set Membership: |
|