Search
Search

LOC102186172| Gene Symbol: LOC102186172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697335.3 | XP_005697392.1 | 1 - 2 | 576 | 80 | |
| GenBank mRNA | JO642304.2 | - | 2 - 3 | 396 | 80 | |
Set Membership: |
|