Search
Search
| Gene Symbol: LPXN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059036.1 | XP_017914525.1 | 3 - 4 | 326 | 79 | |
| XM_018059037.1 | XP_017914526.1 | 3 - 4 | 326 | 79 | ||
| GenBank mRNA | JO579604.1 | - | 3 - 4 | 380 | 79 | |
Set Membership: |
|