Search
Search
| Gene Symbol: CDK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972144.2 | XP_013827598.1 | 4 - 5 | 521 | 87 | |
| XM_013972145.2 | XP_013827599.1 | 3 - 4 | 454 | 87 | ||
| XM_018063772.1 | XP_017919261.1 | 4 - 5 | 518 | 87 | ||
Set Membership: |
|