Search
Search
| Gene Symbol: DCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685022.3 | XP_005685079.1 | 7 - 8 | 1000 | 61 | |
| XM_018053752.1 | XP_017909241.1 | 6 - 7 | 817 | 61 | ||
| GenBank mRNA | JO603787.2 | - | 4 - 5 | 3124 | 61 | |
Set Membership: |
|