Search
Search
| Gene Symbol: APRT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061863.1 | XP_017917352.1 | 4 - 5 | 896 | 68 | |
| GenBank mRNA | JO580041.1 | - | 4 - 5 | 582 | 68 | |
| JO580042.1 | - | 2 - 3 | 923 | 68 | ||
Set Membership: |
|