Search
Search
| Gene Symbol: PRKD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062649.1 | XP_017918138.1 | 10 - 11 | 2321 | 54 | |
| XM_018062650.1 | XP_017918139.1 | 11 - 12 | 2024 | 54 | ||
| XM_018062651.1 | XP_017918140.1 | 9 - 10 | 1305 | 54 | ||
| XM_018062652.1 | XP_017918141.1 | 9 - 10 | 1289 | 54 | ||
| GenBank mRNA | JO425686.1 | - | 8 - 9 | 1359 | 54 | |
| JO579380.1 | - | 8 - 9 | 1372 | 54 | ||
Set Membership: |
|