Search
Search
| Gene Symbol: TNPO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065583.1 | XP_017921072.1 | 11 - 12 | 1290 | 70 | |
| XM_018065584.1 | XP_017921073.1 | 11 - 12 | 1278 | 70 | ||
| XM_018065585.1 | XP_017921074.1 | 11 - 12 | 1274 | 70 | ||
Set Membership: |
|