Search
Search

GOLGB1| Gene Symbol: GOLGB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675002.3 | XP_005675059.2 | 21 - 22 | 9818 | 104 | |
| GenBank mRNA | JO334639.2 | - | 3 - 4 | 334 | 104 | |
| JO348380.2 | - | 1 - 2 | 593 | 104 | ||
| JO372776.2 | - | 1 - 2 | 895 | 104 | ||
| JO446918.2 | - | 2 - 3 | 176 | 104 | ||
| JO548271.2 | - | 2 - 3 | 223 | 104 | ||
| JO603222.2 | - | 2 - 3 | 176 | 104 | ||
| JO640166.2 | - | 1 - 2 | 1387 | 104 | ||
| JO640167.2 | - | 1 - 2 | 1388 | 104 | ||
| JT705791.1 | - | 3 - 4 | 1275 | 104 | ||
| JT717484.1 | - | 2 - 3 | 883 | 104 | ||
| KA340652.1 | - | 1 - 2 | 579 | 104 | ||
Set Membership: |
|