Search
Search

GOLGB1| Gene Symbol: GOLGB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675002.3 | XP_005675059.2 | 9 - 10 | 1429 | 102 | |
| GenBank mRNA | JO334826.2 | - | 1 - 2 | 50 | 102 | |
| JO419873.2 | - | 4 - 5 | 1574 | 102 | ||
| JO430516.2 | - | 4 - 5 | 1506 | 102 | ||
| JO431562.2 | - | 4 - 5 | 1506 | 102 | ||
| JO495891.2 | - | 9 - 10 | 1417 | 102 | ||
| JO611059.2 | - | 1 - 2 | 46 | 102 | ||
| JO611060.2 | - | 1 - 2 | 46 | 102 | ||
| JO616912.2 | - | 4 - 5 | 1472 | 102 | ||
| JT705791.1 | - | 15 - 16 | 9676 | 102 | ||
| JT737680.1 | - | 5 - 6 | 7366 | 102 | ||
| KA314787.1 | - | 4 - 5 | 381 | 102 | ||
Set Membership: |
|