Search
Search

C7H5orf15| Gene Symbol: C7H5orf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682932.3 | XP_005682989.1 | 1 - 2 | 202 | 57 | |
| GenBank mRNA | JO412377.2 | - | 2 - 3 | 1989 | 57 | |
| KA331344.1 | - | 1 - 2 | 179 | 57 | ||
Set Membership: |
|