Search
Search

PTP4A3| Gene Symbol: PTP4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058643.1 | XP_017914132.1 | 3 - 4 | 1160 | 54 | |
| XM_018058644.1 | XP_017914133.1 | 3 - 4 | 1084 | 54 | ||
| XM_018058645.1 | XP_017914134.1 | 3 - 4 | 1052 | 54 | ||
| GenBank mRNA | JO363644.2 | - | 1039 | 54 | ||
Set Membership: |
|