Search
Search

C15H11orf74| Gene Symbol: C15H11orf74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690093.2 | XP_005690150.2 | 2 - 3 | 281 | 99 | |
| XM_005690094.2 | XP_005690151.2 | 3 - 4 | 339 | 99 | ||
| GenBank mRNA | JT722119.1 | - | 2 - 3 | 193 | 99 | |
| KA326007.1 | - | 2 - 3 | 271 | 99 | ||
Set Membership: |
|