Search
Search

CIAO2B| Gene Symbol: CIAO2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692114.3 | XP_005692171.2 | 3 - 4 | 471 | 79 | |
| GenBank mRNA | JO373818.2 | - | 2 - 3 | 261 | 79 | |
| JO451787.2 | - | 3 - 4 | 407 | 79 | ||
Set Membership: |
|