Search
Search

LOC102172109| Gene Symbol: LOC102172109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039154.1 | XP_017894643.1 | 2 - 3 | 303 | 77 | |
| GenBank mRNA | JO643491.2 | - | 3 - 4 | 557 | 77 | |
Set Membership: |
|