Search
Search
| Gene Symbol: MYH10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064633.1 | XP_017920122.1 | 22 - 23 | 2796 | 53 | |
| XM_018064634.1 | XP_017920123.1 | 22 - 23 | 2917 | 53 | ||
| XM_018064635.1 | XP_017920124.1 | 22 - 23 | 2797 | 53 | ||
| XM_018064636.1 | XP_017920125.1 | 21 - 22 | 2765 | 53 | ||
| XM_018064637.1 | XP_017920126.1 | 21 - 22 | 2731 | 53 | ||
| XM_018064638.1 | XP_017920127.1 | 20 - 21 | 2700 | 53 | ||
| GenBank mRNA | JO632898.2 | - | 2126 | 53 | ||
Set Membership: |
|