Search
Search
| Gene Symbol: HAGH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040692.1 | XP_017896181.1 | 7 - 8 | 797 | 57 | |
| GenBank mRNA | JO427857.1 | - | 2 - 3 | 1814 | 57 | |
| JO634362.2 | - | 2 - 3 | 669 | 57 | ||
Set Membership: |
|