Search
Search
| Gene Symbol: MTX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685087.3 | XP_005685144.1 | 5 - 6 | 523 | 91 | |
| XM_005685089.3 | XP_005685146.1 | 5 - 6 | 523 | 91 | ||
| XM_013966735.2 | XP_013822189.1 | 5 - 6 | 523 | 91 | ||
| XM_013966736.2 | XP_013822190.1 | 5 - 6 | 523 | 91 | ||
| GenBank mRNA | JT716218.1 | - | 5 - 6 | 604 | 91 | |
Set Membership: |
|