Search
Search

C18H16orf70| Gene Symbol: C18H16orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692120.3 | XP_005692177.1 | 15 - 16 | 1242 | 99 | |
| XM_005692121.3 | XP_005692178.1 | 15 - 16 | 1228 | 99 | ||
| XM_018062103.1 | XP_017917592.1 | 15 - 16 | 1230 | 99 | ||
Set Membership: |
|