Search
Search
| Gene Symbol: BAG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038758.1 | XP_017894247.1 | 10 - 11 | 1541 | 62 | |
| XM_018038759.1 | XP_017894248.1 | 10 - 11 | 1641 | 62 | ||
| XM_018038760.1 | XP_017894249.1 | 10 - 11 | 1403 | 62 | ||
| XM_018038761.1 | XP_017894250.1 | 10 - 11 | 1541 | 62 | ||
| XM_018038762.1 | XP_017894251.1 | 10 - 11 | 1685 | 62 | ||
| XM_018038763.1 | XP_017894252.1 | 10 - 11 | 1542 | 62 | ||
| XM_018038764.1 | XP_017894253.1 | 10 - 11 | 1541 | 62 | ||
Set Membership: |
|