Search
Search

ZNF654| Gene Symbol: ZNF654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054696.1 | XP_017910185.1 | 2 - 3 | 437 | 78 | |
| XM_018054719.1 | XP_017910208.1 | 2 - 3 | 436 | 78 | ||
| GenBank mRNA | JO422322.2 | - | 2 - 3 | 225 | 78 | |
| JO441715.2 | - | 2 - 3 | 433 | 78 | ||
| JO444692.2 | - | 2 - 3 | 433 | 78 | ||
| JO601482.2 | - | 2 - 3 | 461 | 78 | ||
| JT705486.1 | - | 2 - 3 | 753 | 78 | ||
Set Membership: |
|