Search
Search
| Gene Symbol: AADAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683509.3 | XP_005683566.1 | 12 - 13 | 1550 | 103 | |
| XM_013965831.2 | XP_013821285.1 | 11 - 12 | 1484 | 103 | ||
Set Membership: |
|